Poly(methylhydrosiloxane)
Poly(methylhydrosiloxane)
Poly(methylhydrosiloxane)
Poly(methylhydrosiloxane)
Poly(methylhydrosiloxane)
Thermo Scientific Chemicals
Poly(methylhydrosiloxane)
CAS: 63148-57-2 | (CH4OSi)n
Have Questions?
Catalog NumberQuantity
ALFL14561.1850 g
Catalog number ALFL14561.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(methylhydrosiloxane)
CAS63148-57-2
Recommended StorageAmbient temperatures
TSCAYes
Molecular Formula(CH4OSi)n
View more
Poly(methylhydrosiloxane) is used in the reduction of esters to alcohols, catalyzed by a combination of titanocene dichloride and either n-BuLi or EtMgBr. It is a safer alternative to triethoxysilane, B22063, for reduction of phosphine oxides to phosphines, catalyzed by Ti(O-i-Pr)4. It is also used in greener amine synthesis by reductive amination as an alternative to borohydrides.