Kerosene, low odor
Thermo Scientific Chemicals
Kerosene, low odor
CAS: 64742-47-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALFL14479.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialKerosene
Name Notelow odor
CAS64742-47-8
Health Hazard 1H227-H304
Health Hazard 2GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
View more
Kerosene is ideal for dilution of oils and other organic fluids for determination of metal traces. It is used in paints & coatings, consumer products, printing inks and agricultural chemical applications. It is also used in food contact applications such as aluminum rolling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Kerosene is ideal for dilution of oils and other organic fluids for determination of metal traces. It is used in paints & coatings, consumer products, printing inks and agricultural chemical applications. It is also used in food contact applications such as aluminum rolling.
Solubility
Soluble in water (0.02 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only
General References:
  1. Philippe Dagaut. On the kinetics of hydrocarbons oxidation from natural gas to kerosene and diesel fuel. Phys. Chem. Chem. Phys. 2002, 4 2079-2094.
  2. M.L. Jeswani. Effect of the addition of graphite powder to kerosene used as the dielectric fluid in electrical discharge machining. Wear. 1981, 70 (2), 133-139.