Amberlite™ IRC-120(H), ion exchange resin
Amberlite™ IRC-120(H), ion exchange resin
Amberlite™ IRC-120(H), ion exchange resin
Amberlite™ IRC-120(H), ion exchange resin
Thermo Scientific Chemicals
Amberlite™ IRC-120(H), ion exchange resin
CAS: 78922-04-0 | C13H10ClNO4S | 311.736 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFL14285.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ IR-120(H)
Name Noteion exchange resin
CAS78922-04-0
Health Hazard 1H318
Health Hazard 2GHS H Statement
H318
Causes serious eye damage.
View more
Amberlite™ IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Amberlite|r IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Naushad, M.; ALOthman, Z. A.; Sharma, G.; Inamuddin. Kinetics, isotherm and thermodynamic investigations for the adsorption of Co(II) ion onto crystal violet modified amberlite IR-120 resin Ionics. 2015, 21 (5), 1453-1459.
  2. Kamal, A.; Babu, K. S.; Hussaini, S. M. A.; Mahesh, R.; Alarifi, A. Amberlite IR-120H, an efficient and recyclable solid phase catalyst for the synthesis of quinoxalines: a greener approach. Tetrahedron Lett. 2015, 56 (21), 2803-2808.