DL-Lactic acid, 80-85% aq. soln.
DL-Lactic acid, 80-85% aq. soln.
DL-Lactic acid, 80-85% aq. soln.
DL-Lactic acid, 80-85% aq. soln.
Thermo Scientific Chemicals
DL-Lactic acid, 80-85% aq. soln.
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14259.0B1000 g
Catalog number ALFL14259.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Lactic acid
CAS50-21-5
Boiling Point122°C (15 mmHg)
Health Hazard 1H315-H318
Health Hazard 2GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
View more
DL-Lactic acid is used to produce serial products or widely used in food, vintage, beverage, drugs, polymerization, textile, leather, tobacco, feed, plastic chemicals, pesticide, polymer solution and other industry. Lactic acid is also used as an acidifying agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water and in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Colin G. Pitt; Margit M. Gratzl; A. Robert Jeffcoat; Ruth Zweidinger and Anton Schindler. Sustained drug delivery systems II: Factors affecting release rates from poly(ε-caprolactone) and related biodegradable polyesters. Journal of Pharmaceutical Sciences. 1979, 68, (12), 1534-1538.
  2. Schakenraad, J.M.; Nieuwenhuis, P.; Molenaar, I.; Helder, Janneke; Dijkstra, P.J. and Feijen, J. In vivo and in vitro degradation of glycine/DL-lactic acid copolymers. Journal of Biomedical Materials Research. 1979, 23, (11), 1271-1288.