9-Bromo-9-phenylfluorene, 96%
9-Bromo-9-phenylfluorene, 96%
9-Bromo-9-phenylfluorene, 96%
9-Bromo-9-phenylfluorene, 96%
Thermo Scientific Chemicals
9-Bromo-9-phenylfluorene, 96%
CAS: 55135-66-5 | C19H13Br | 321.217 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14225.065 g
Catalog number ALFL14225.06
Price (MYR)
1,123.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,123.00
EA
Add to cart
Specifications
Chemical Name or Material9-Bromo-9-phenylfluorene
CAS55135-66-5
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c