tert-Butyl methyl ether, 99%
tert-Butyl methyl ether, 99%
tert-Butyl methyl ether, 99%
tert-Butyl methyl ether, 99%
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14030.AU1000 mL
Catalog number ALFL14030.AU
Price (MYR)
356.00
EA
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (MYR)
356.00
EA
Add to cart
Specifications
Chemical Name or Materialtert-Butyl methyl ether
Melting Point-109°C
CAS1634-04-4
Health Hazard 1H225-H315-H335
Health Hazard 2GHS H Statement
H225-H315
Highly flammable liquid and vapor.
Causes skin irritation.
View more
Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).
Solubility
Miscible with ethanol and ethyl ether. Immiscible with water.
Notes
Store in a cool, dry, well-ventilated area. It is hygroscopic in nature.
RUO – Research Use Only
General References:
  1. Fischer, A.; Oehm, C.; Selle, M.; Werner, P.; Oehm; Selle; Werner. Biotic and abiotic transformations of methyl tertiary butyl ether (MTBE). Environ. Sci. Pollut. Res. Int. 2005, 12 (6), 381-389.
  2. Matyash, V.; Liebisch, G.; Kurzchalia, T. V.; Shevchenko, A.; Schwudke, D. Lipid extraction by methyl-tert-butyl ether for high-throughput lipidomics. J. Lipid Res. 2008, 49 (5), 1137-1146.