Silica gel 60, 0.036-0.071mm (215-400 mesh)
Silica gel 60, 0.036-0.071mm (215-400 mesh)
Silica gel 60, 0.036-0.071mm (215-400 mesh)
Silica gel 60, 0.036-0.071mm (215-400 mesh)
Thermo Scientific Chemicals
Silica gel 60, 0.036-0.071mm (215-400 mesh)
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1000 g
2500 g
Catalog number ALFL14004.36
Price (MYR)
463.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
463.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilica gel 60
Name Note0.036 to 0.071mm
CAS7631-86-9
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
View more
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
Solubility
Soluble in hydrofluoric acid. Insoluble in water.
Notes
Keep the container tightly sealed. Store in cool, dry conditions in a well sealed container. Incompatible with hydrogen fluoride.
RUO – Research Use Only
General References:
  1. Suitable for conventional and flash column chromatography.
  2. Khatry, S.; Sood, N.; Arora, S. Surface solid dispersion-a review. Int. J. Pharm. Sci. Nanotech. 2013, 6 (1), 1915-24.
  3. Ray, D.; Mittal, N.; Chung, W. J. Phosphorous pentoxide mediated synthesis of 5-HMF in ionic liquid at low temperature. Carbohydr. Res. 2011, 346 (14), 2145-2148.