N-Methylaniline, 98%
N-Methylaniline, 98%
N-Methylaniline, 98%
N-Methylaniline, 98%
Thermo Scientific Chemicals
N-Methylaniline, 98%
CAS: 100-61-8 | C7H9N | 107.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13873.36500 g
Catalog number ALFL13873.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Methylaniline
CAS100-61-8
Health Hazard 1H227-H301+H311+H331-H373
Health Hazard 2GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c
View more
N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
RUO – Research Use Only
General References:
  1. N-Alkylanilines react with triethanolamine in dioxan (autoclave) in the presence of Dichlorotris(triphenyl phosphine) ruthenium(II) , L00373, to give the corresponding 1-alkylindole in good yield: Synth. Commun., 26, 1349 (1996).
  2. Emel’yanenko, V. N.; Pimerzin, A. A.; Turovtsev, V. V.; Verevkin, S. P. Benchmark Thermochemistry of N-Methylaniline. J. Phys. Chem. A 2015, 119 (10), 2142-2152.
  3. Belyaeva, E. I.; Zrelova, L. V.; Marchenko, D. Y.; Dedov, A. G. Colorimetric determination of N-methylaniline in hydrocarbon media. Petro. Chem. 2015, 55 (1), 74-79.