N-Methylcyclohexylamine, 98%
N-Methylcyclohexylamine, 98%
N-Methylcyclohexylamine, 98%
N-Methylcyclohexylamine, 98%
Thermo Scientific Chemicals
N-Methylcyclohexylamine, 98%
CAS: 100-60-7 | C7H15N | 113.20 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13872.AC25 mL
Catalog number ALFL13872.AC
Price (MYR)
238.00
EA
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
238.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Methylcyclohexylamine
CAS100-60-7
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301-H314-H318-H226-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Health Hazard 3GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.