Isophorone diisocyanate, mixture of isomers, 98%
Isophorone diisocyanate, mixture of isomers, 98%
Isophorone diisocyanate, mixture of isomers, 98%
Isophorone diisocyanate, mixture of isomers, 98%
Isophorone diisocyanate, mixture of isomers, 98%
Thermo Scientific Chemicals
Isophorone diisocyanate, mixture of isomers, 98%
CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Catalog number ALFL13759.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIsophorone Diisocyanate
Name Notemixture of isomers
CAS4098-71-9
Health Hazard 1H315-H317-H319-H330-H334-H335
Health Hazard 2GHS H Statement
H330-H334-H315-H319-H317-H335-H204
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.
View more
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.
Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
RUO – Research Use Only
General References:
  1. Vincent, B. J.; Natarajan, B. Kinetics of thermal degradation of water borne polyurethane dispersion containing polycaprolactone with either isophorone diisocyanate or metatetramethyl xylene diisocyanate. J. Therm. Anal. Calorim. 2015, 119 (2), 1373-1379.
  2. Dzalmukhanova, A. S.; Lodygina, V. P.; Komratova, V. V.; Karpov, S. V.; Malkov, G. V.; Badamshina, E. R. IR Spectroscopy Method for Determining The Reactivity of Isocyanate Groups in Isophorone Diisocyanate Reactions. J. Appl. Spectrosc. 2015, 82 (1), 145-148.