2-Amino-5-n-propylsulfonylbenzimidazole, 98+%
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%
Thermo Scientific Chemicals
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%
CAS: 80983-34-2 | C10H13N3O2S | 239.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number ALFL13711.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-n-propylsulfonylbenzimidazole
CAS80983-34-2
Melting Point220°C to 222°C
Recommended StorageAmbient temperatures
TSCANo