Polysorbate 80
Polysorbate 80
Polysorbate 80
Thermo Scientific Chemicals
Polysorbate 80
CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13315.AE100 mL
Catalog number ALFL13315.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPolysorbate 80
CAS9005-65-6
Boiling Point>100°C
Recommended StorageAmbient temperatures
RTECS NumberWG2932500
View more
Used for solubilizing membrane proteins during isolation and purification.Polysorbate 80 is used as a surfactant in soaps, cosmetics, body washes, hair shampoos, hair loss treatments, lotions, creams, bath oils and in mouthwash in the health care industry. It serves as an emulsifier in foods such as ice cream and in the preparation of antiarrhythmic amiodarone. Further, it acts as a solubilizing agent and excipient in the pharmaceutical industry.