2-Pentanone, 99+%
2-Pentanone, 99+%
2-Pentanone, 99+%
Thermo Scientific Chemicals
2-Pentanone, 99+%
CAS: 107-87-9 | C5H10O | 86.134 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13262.22100 g
Catalog number ALFL13262.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Pentanone
CAS107-87-9
Health Hazard 1H225-H302-H319
Health Hazard 2GHS H Statement
H225-H302-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
View more
2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Kang, J.; Vonderheide, A.; Guliants, V. V. Deuterium-Labeling Study of the Hydrogenation of 2-Methylfuran and 2,5-Dimethylfuran over Carbon-Supported Noble Metal Catalysts. ChemSusChem 2015, 8 (18), 3044-3047.
  2. Ye, M.; Chien, P. J.; Toma, K.; Arakawa, T.; Mitsubayashi, K. An acetone bio-sniffer (gas phase biosensor) enabling assessment of lipid metabolism from exhaled breath. Biosens. Bioelectron. 2015, 73, 208-213.