L-Lactic acid, anhydrous, 98%
L-Lactic acid, anhydrous, 98%
L-Lactic acid, anhydrous, 98%
L-Lactic acid, anhydrous, 98%
Thermo Scientific Chemicals
L-Lactic acid, anhydrous, 98%
CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFL13242.22
Price (MYR)
4,992.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
4,992.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Lactic acid
Name Noteanhydrous
CAS79-33-4
Health Hazard 1H315-H318
Health Hazard 2GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
View more
Useful chiral synthon, building block for depsipeptides.L-Lactic acid is an intermediate in the fermentation of sugar. It plays a vital role in several biochemical processes. It is a major ingredient in processed foods such as yogurt, cottage cheeses, koumiss and kombucha. It gives the sour flavor to sourdough breads. It finds application in pharmaceuticals to prepare water-soluble lactates, topical preparations and cosmetics. It acts as a food preservative, flavoring agent, curing agent, descaler, soap-scum remover and an anti-bacterial agent. It is an active component of lactated Ringer's solution and Hartmann's solution. Further, it is used for fluid resuscitation after blood loss due to trauma and surgery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Useful chiral synthon, building block for depsipeptides.L-Lactic acid is an intermediate in the fermentation of sugar. It plays a vital role in several biochemical processes. It is a major ingredient in processed foods such as yogurt, cottage cheeses, koumiss and kombucha. It gives the sour flavor to sourdough breads. It finds application in pharmaceuticals to prepare water-soluble lactates, topical preparations and cosmetics. It acts as a food preservative, flavoring agent, curing agent, descaler, soap-scum remover and an anti-bacterial agent. It is an active component of lactated Ringer′s solution and Hartmann′s solution. Further, it is used for fluid resuscitation after blood loss due to trauma and surgery.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong bases, strong oxidizing agents, strong reducing agents and metals.
RUO – Research Use Only
General References:
  1. Lv, X.; Guo, Y.; Zhuang, Y.; Wang, Y. Optimization and validation of an extraction method and HPAEC-PAD for determination of residual sugar composition in l-lactic acid industrial fermentation broth with a high salt content. Anal. Methods 2015, 7 (21), 9076-9083.
  2. Yamada, S.; Takasu, A.; Takayama, S.; Kawamura, K. Microwave-assisted solution polycondensation of L-lactic acid using a Dean-Stark apparatus for a non-thermal microwave polymerization effect induced by the electric field. Polym. Chem. 2014, 5 (18), 5283-5288.