2-Butanone, 99%
2-Butanone, 99%
2-Butanone, 99%
Thermo Scientific Chemicals
2-Butanone, 99%
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13185.0F2500 mL
Catalog number ALFL13185.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Specifications
Chemical Name or Material2-Butanone
Melting Point-87°C
CAS78-93-3
Health Hazard 1H225-H319-H336-H373
Health Hazard 2GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
View more
2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.
Solubility
Miscible with cold water, diethyl ether and acetone.
Notes
It is stable, incompatible with oxidizing agents, bases, strong reducing agents.
RUO – Research Use Only
General References:
  1. Suzuki, T.; Morita, K.; Tsuchida, M.; Hiroi, K. Iridium-Catalyzed Oppenauer Oxidations of Primary Alcohols Using Acetone or 2-Butanone as Oxidant. J. Org. Chem. 2003, 68 (4), 1601-1602.
  2. Szollosi, G.; Bartok, M. Catalytic transfer hydrogenation of 2-butanone on MgO. New active surface sites generated by treatment with chloroform. Catal. Lett. 1999, 59 (2-4), 179-185.