6-Heptenoic acid, 96%
6-Heptenoic acid, 96%
6-Heptenoic acid, 96%
6-Heptenoic acid, 96%
Thermo Scientific Chemicals
6-Heptenoic acid, 96%
CAS: 1119-60-4 | C7H12O2 | 128.171 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13144.031 g
Catalog number ALFL13144.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material6-Heptenoic acid
CAS1119-60-4
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
General References:
  1. Nisha; Kewal, K.; Vipan, K. Prodigiosin alkaloids: recent advancements in total synthesis and their biological potential. RSC Adv. 2015, 5 (15), 10899-10920.
  2. Wei, S.; Li, Y.; Ma, Q.; Lou, S.; Schreiner, M. Characterization of Tang Dynasty lamp oil remains by using pyrolysis gas chromatography and mass spectrometry. J. Anal. Appl. Pyrolysis 2015, 116, 237-242.