Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol
Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol
Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol
Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol
Thermo Scientific Chemicals
Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol
CAS: 106-63-8 | C7H12O2 | 128.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12739.AE100 mL
Catalog number ALFL12739.AE
Price (MYR)
329.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
329.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsobutyl acrylate
Name Notestabilized with 100ppm 4-methoxyphenol
CAS106-63-8
Health Hazard 1H226-H312+H332-H315-H317-H319
Health Hazard 2GHS H Statement
H226-H312-H332-H315-H317
Flammable liquid and vapor.