4-(Trifluoromethyl)phenyl isocyanate, 98%
4-(Trifluoromethyl)phenyl isocyanate, 98%
4-(Trifluoromethyl)phenyl isocyanate, 98%
4-(Trifluoromethyl)phenyl isocyanate, 98%
Thermo Scientific Chemicals
4-(Trifluoromethyl)phenyl isocyanate, 98%
CAS: 1548-13-6 | C8H4F3NO | 187.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12576.065 g
Catalog number ALFL12576.06
Price (MYR)
914.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
914.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)phenyl isocyanate
CAS1548-13-6
Health Hazard 1H227-H302+H312-H315-H319-H330-H334-H335
Health Hazard 2GHS H Statement
H330-H334-H227-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P284-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
View more
4-(Trifluoromethyl)phenyl isocyanate has been used in the synthesis of fluorescent photoinduced electron transfer (PET) chemosensors and 2-ethyl-6-[(4-trifluoromethylphenylcarbamoyl)hydrazino]-benzo[de]iso-quinoline-1,3-dione, an amidourea-based sensor for anions. It has also been used in the synthesis of 1,8-naphthalimide based colorimetric and fluorescent sensor for anions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethyl)phenyl isocyanate has been used in the synthesis of fluorescent photoinduced electron transfer (PET) chemosensors and 2-ethyl-6-[(4-trifluoromethylphenylcarbamoyl)hydrazino]-benzo[de]iso-quinoline-1,3-dione, an amidourea-based sensor for anions. It has also been used in the synthesis of 1,8-naphthalimide based colorimetric and fluorescent sensor for anions.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
RUO – Research Use Only
General References:
  1. Thorfinnur Gunnlaugsson; Anthony P.Davis; John E O'Brien; Mark Glynn. Synthesis and photophysical evaluation of charge neutral thiourea or urea based fluorescent PET sensors for bis-carboxylates and pyrophosphate. Organic & Biomolecular Chemistry. 2005, 3,(1), 48-56.
  2. Thorfinnur Gunnlaugsson; Paul E.Kruger; Paul Jensen; Juliann Tierney; Haslin Dato Paduka Ali; Gillian M.Hussey. Colorimetric naked eye sensing of anions in aqueous solution. J. Org. Chem. 2005, 70,(26), 10875-10878.