tert-Butyl alcohol, 99%
tert-Butyl alcohol, 99%
tert-Butyl alcohol, 99%
tert-Butyl alcohol, 99%
Thermo Scientific Chemicals
tert-Butyl alcohol, 99%
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12338.AK250 mL
Catalog number ALFL12338.AK
Price (MYR)
284.00
EA
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
284.00
EA
Add to cart
Specifications
Chemical Name or Materialtert-Butyl alcohol
CAS75-65-0
Health Hazard 1H225-H319-H332-H335-H336
Health Hazard 2GHS H Statement
H225-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
View more
tert-Butyl alcohol is used as a solvent and ethanol denaturant. It is an active ingredient of paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl alcohol is used as a solvent and ethanol denaturant. It is an active ingredient of paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.
Solubility
Miscible with water, ethanol, ethyl ether and chloroform.
Notes
Incompatible with strong oxidizing agents, copper, alkali metals and aluminum.
RUO – Research Use Only
General References:
  1. Munjal, B.; Bansal, A. K. Impact of Tert-Butyl Alcohol on Crystallization Kinetics of Gemcitabine Hydrochloride in Frozen Aqueous Solutions. J. Pharm. Sci. 2015, 104 (1), 87-97.
  2. Danaiah, P.; Ravi Kumar, P.; Rao, Y. V. H. Performance and emission prediction of a tert butyl alcohol gasoline blended spark-ignition engine using artificial neural networks. Int. J. Ambient Energy 2015, 36 (1), 31-39.