2-n-Hexyldecanoic acid, 97%
2-n-Hexyldecanoic acid, 97%
2-n-Hexyldecanoic acid, 97%
2-n-Hexyldecanoic acid, 97%
Thermo Scientific Chemicals
2-n-Hexyldecanoic acid, 97%
CAS: 25354-97-6 | C16H32O2 | 256.43 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12238.1425 g
Catalog number ALFL12238.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-n-Hexyldecanoic acid
CAS25354-97-6
Melting Point10°C
Recommended StorageAmbient temperatures
Density0.88