Bis(2-benzamidophenyl) disulfide, 97%
Bis(2-benzamidophenyl) disulfide, 97%
Bis(2-benzamidophenyl) disulfide, 97%
Bis(2-benzamidophenyl) disulfide, 97%
Thermo Scientific Chemicals
Bis(2-benzamidophenyl) disulfide, 97%
CAS: 135-57-9 | C26H20N2O2S2 | 456.578 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12110.1425 g
Catalog number ALFL12110.14
Price (MYR)
273.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
273.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis(2-benzamidophenyl) disulfide
CAS135-57-9
Melting Point140°C to 143°C
Recommended StorageAmbient temperatures
RTECS NumberCV8700000