1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
Thermo Scientific Chemicals
1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
CAS: 1200-27-7 | C10H14FN | 167.227 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11858.065 g
Catalog number ALFL11858.06
Price (MYR)
2,515.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,515.00
EA
Add to cart
Specifications
Chemical Name or Material1-(4-Fluorophenyl)-2-methyl-2-propylamine
CAS1200-27-7
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c