2-Ethyl-1-butene, 97%
2-Ethyl-1-butene, 97%
2-Ethyl-1-butene, 97%
2-Ethyl-1-butene, 97%
Thermo Scientific Chemicals
2-Ethyl-1-butene, 97%
CAS: 760-21-4 | C6H12 | 84.162 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11553.065 g
Catalog number ALFL11553.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Ethyl-1-butene
CAS760-21-4
Health Hazard 1H225-H315-H319-H335
Health Hazard 2GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
View more
2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Soluble in alcohol, and methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only
General References:
  1. I. Kind.; T. Berndt.; O. Böge. Gas-phase rate constants for the reaction of NO3 radicals with a series of cyclic alkenes, 2-ethyl-1-butene and 2,3-dimethyl-1,3-butadiene. Chemical Physics Letters. 1998, 288 (1), 111-118.
  2. Hun-Soo Byun.; Tae-Hyun Choi. Vapor-Liquid equilibria measurement of carbon dioxide+1-hexene and carbon dioxide+2-ethyl-1-butene systems at high pressure. Korean Journal of Chemical Engineering. 2004, 21 (5), 1032-1037.