Sorbitan monostearate
Sorbitan monostearate
Sorbitan monostearate
Sorbitan monostearate
Thermo Scientific Chemicals
Sorbitan monostearate
CAS: 1338-41-6 | C24H46O6 | 430.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFL11435.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSorbitan monostearate
CAS1338-41-6
Melting Point53°C to 57°C
Recommended StorageAmbient temperatures
Density1
View more
Sorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.
Solubility
Soluble in ethanol (50 mg/ml), isopropanol, mineral oil and vegetable oil. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Sudaxshina Murdan.; Gregory Gregoriadis.; Alexander T. Florence.Novel sorbitan monostearate organogels. Journal of Pharmaceutical Sciences. 1999, 88 (6), 608-614.
  2. Elisabetta Gianasi.; Fausto Cociancich.; Ijeoma F. Uchegbu.; Alexander T. Florence.; Ruth Duncan. Pharmaceutical and biological characterisation of a doxorubicin-polymer conjugate (PK1) entrapped in sorbitan monostearate Span 60 niosomes. International Journal of Pharmaceutics. 1997, 148 (2), 139-148.
  3. Span™ is a registered trademark of ICI Group.
  4. Nonionic surfactant.