1-Hydroxycyclohexyl phenyl ketone, 98%
1-Hydroxycyclohexyl phenyl ketone, 98%
1-Hydroxycyclohexyl phenyl ketone, 98%
1-Hydroxycyclohexyl phenyl ketone, 98%
Thermo Scientific Chemicals
1-Hydroxycyclohexyl phenyl ketone, 98%
CAS: 947-19-3 | C13H16O2 | 204.27 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11377.36500 g
Catalog number ALFL11377.36
Price (MYR)
2,028.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,028.00
EA
Add to cart
Specifications
Chemical Name or Material1-Hydroxycyclohexyl phenyl ketone
CAS947-19-3
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
View more
1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.
Solubility
Slightly soluble in water (1108 mg/L at 25°C). Soluble in acetone, butyl acetate, methanol and toluene.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only