(Bromomethyl)cyclohexane, 98%
(Bromomethyl)cyclohexane, 98%
(Bromomethyl)cyclohexane, 98%
(Bromomethyl)cyclohexane, 98%
Thermo Scientific Chemicals
(Bromomethyl)cyclohexane, 98%
CAS: 2550-36-9 | C7H13Br | 177.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11343.065 g
Catalog number ALFL11343.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material(Bromomethyl)cyclohexane
CAS2550-36-9
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
View more
(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Philip Boudjouk.; Ratnasabapathy Sooriyakumaran.; Craig A. Kapfer. The reaction of magnesium with cis-1,3,5-tris(bromomethyl)cyclohexane. Evidence for a soluble tri-Grignard. Journal of Organometallic Chemistry. 1985, 281 (2-3), 21-23.
  2. George P Luke.; Dennis A Holt. Synthesis of (S)-5-(1-aminoethyl)-2-(cyclohexylmethoxy)benzamide. Tetrahedron: Asymmetry. 1999, 10 (22), 4393-4403.