Chromotrope 2B
Chromotrope 2B
Chromotrope 2B
Thermo Scientific Chemicals
Chromotrope 2B
CAS: 548-80-1 | C16H9N3Na2O10S2 | 513.36 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11330.1425 g
Catalog number ALFL11330.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialChromotrope 2B
Melting Point>300°C
CAS548-80-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c