View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Acetyl-D-mannosamine monohydrate
CAS1071625-31-4
Melting Point∼130°C (decomposition)
Recommended StorageKeep cold
TSCAYes