4-Bromophenyl isothiocyanate, 97%
4-Bromophenyl isothiocyanate, 97%
4-Bromophenyl isothiocyanate, 97%
4-Bromophenyl isothiocyanate, 97%
Thermo Scientific Chemicals
4-Bromophenyl isothiocyanate, 97%
CAS: 1985-12-2 | C7H4BrNS | 214.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11101.065 g
Catalog number ALFL11101.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromophenyl isothiocyanate
CAS1985-12-2
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c