1-Octadecene, tech. 90%
1-Octadecene, tech. 90%
1-Octadecene, tech. 90%
1-Octadecene, tech. 90%
1-Octadecene, tech. 90%
Thermo Scientific Chemicals
1-Octadecene, tech. 90%
CAS: 112-88-9 | C18H36 | 252.486 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number ALFL11004.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Octadecene
CAS112-88-9
Health Hazard 1H304
Health Hazard 3P301+P310-P331-P501c
Melting Point15°C to 17°C
View more
1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.
Solubility
Miscible with alcohol, acetone and ether. Immiscible with water.
Notes
Avoid reaction with strong Lewis or mineral acids. Free radical initiators should be avoided. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Sanoria, A.; Schuster, T.; Brull, R. Raman spectroscopy as a non-destructive tool to quantify the comonomer content in ethylene/1-olefin copolymers. Anal. Methods 2015, 7 (12), 5245-5253.
  2. Moya, C.; Morales, M. P.; Batlle, X.; Labarta, A. Tuning the magnetic properties of Co-ferrite nanoparticles through the 1,2-hexadecanediol concentration in the reaction mixture. Phys. Chem. Chem. Phys. 2015, 17 (19), 13143-13149.