Vitamin K{1}
Vitamin K{1}
Vitamin K{1}
Thermo Scientific Chemicals
Vitamin K{1}
Important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver. | CAS: 84-80-0 | C31H46O2 | 450.707 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL10575.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialVitamin K1
CAS84-80-0
Melting Point∼-20°C
Recommended StorageKeep cold
Density0.984
View more
Vitamin K{1} is an important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver.