Thiophene-2,5-dicarboxylic acid, 97%
Thiophene-2,5-dicarboxylic acid, 97%
Thiophene-2,5-dicarboxylic acid, 97%
Thiophene-2,5-dicarboxylic acid, 97%
Thermo Scientific Chemicals
Thiophene-2,5-dicarboxylic acid, 97%
CAS: 4282-31-9 | C6H4O4S | 172.154 g/mol
Have Questions?
Catalog NumberQuantity
ALFL10293.065 g
Catalog number ALFL10293.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThiophene-2,5-dicarboxylic acid
CAS4282-31-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Bang-Lin Chen; Kum-Fun Mok ; Siu-Choon Ng; Michael G.B.Drew. Syntheses, structures and properties of copper(II) complexes with thiophene-2,5-dicarboxylic acid (H2Tda) and nitrogen-containing ligands. Polyhedron. 1999, 18, (8-9),1211-1220
  2. P.Guerriero; U.Casellato; S.Sitran; P.A.Vigato; R.Graziani. Preparation and properties of lanthanide complexes with pyridine-2,6-dicarboxylic and thiophene-2,5-dicarboxylic acids. Crystal structure of diaqua-bis(dipicolinate)lanthanum(III) tetrahydrate. Inorganica Chimica Acta. 1987, 133, (2),337-345