2-Mercaptopropionic acid, 97%
2-Mercaptopropionic acid, 97%
2-Mercaptopropionic acid, 97%
2-Mercaptopropionic acid, 97%
Thermo Scientific Chemicals
2-Mercaptopropionic acid, 97%
CAS: 79-42-5 | C3H6O2S | 106.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFL10257.36500 g
Catalog number ALFL10257.36
Price (MYR)
783.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
783.00
EA
Add to cart
Specifications
Chemical Name or Material2-Mercaptopropionic acid
CAS79-42-5
Health Hazard 1H227-H290-H302-H314-H335
Health Hazard 2GHS H Statement
H301-H331-H314-H318-H227
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3P210-P234-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c