4-Nitrophenyl-beta-D-glucopyranoside, 98+%
4-Nitrophenyl-beta-D-glucopyranoside, 98+%
4-Nitrophenyl-beta-D-glucopyranoside, 98+%
4-Nitrophenyl-beta-D-glucopyranoside, 98+%
Thermo Scientific Chemicals
4-Nitrophenyl-beta-D-glucopyranoside, 98+%
CAS: 2492-87-7 | C12H15NO8 | 301.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL10154.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl-beta-D-glucopyranoside
CAS2492-87-7
Melting Point166°C to 167°C
Recommended StorageStore at -20°C
EINECS Number219-661-3
View more
4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.
Solubility
Soluble in water, methanol, warm ethanol, dimethyl sulfoxide and dimethylformamide. Insoluble in ether.
Notes
Store in a cool place. Incompatible with strong bases.
RUO – Research Use Only
General References:
  1. Substrate for ß -glucosidase: J. Biol. Chem., 255, 9030 (1980).
  2. Byun, D. H.; Choi, H. J.; Lee, H. W.; Jeon, H. Y.; Choung, W. J.; Shim, J. H. Properties and applications of beta-glycosidase from Bacteroides thetaiotaomicron that specifically hydrolyses isoflavone glycosides. Int. J. Food Sci. Technol. 2015, 50 (6), 1405-1412.
  3. Hofer, C. J.; Zlateski, V.; Stoessel, P. R.; Paunescu, D.; Schneider, E. M.; Grass, R. N.; Zeltner, M.; Stark, W. J. Stable dispersions of azide functionalized ferromagnetic metal nanoparticles. Chem. Commun. 2015, 51 (10), 1826-1829.