4-Nitrophenyl-alpha-D-glucopyranoside, 98+%
4-Nitrophenyl-alpha-D-glucopyranoside, 98+%
4-Nitrophenyl-alpha-D-glucopyranoside, 98+%
4-Nitrophenyl-alpha-D-glucopyranoside, 98+%
Thermo Scientific Chemicals
4-Nitrophenyl-alpha-D-glucopyranoside, 98+%
CAS: 3767-28-0 | C12H15NO8 | 301.251 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL10070.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl-alpha-D-glucopyranoside
CAS3767-28-0
Melting Point206°C to 207°C
Recommended StorageKeep cold
EINECS Number223-189-3
View more
4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.
Solubility
Soluble in water, warm ethanol and methanol.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Substrate for ɑ -glucosidase: Anal. Biochem., 113, 188 (1981).
  2. Zhao, D. G.; Zhou, A. Y.; Du, Z.; Zhang, Y.; Zhang, K.; Ma, Y. Y. Coumarins with alpha-glucosidase and alpha-amylase inhibitory activities from the flower of Edgeworthia gardneri. Fitoterapia 2015, 107, 122-127.
  3. Madeira, P. P.; Bessa, A.; Loureiro, J. A.; Álvares-Ribeiro, L.; Rodrigues, A. E.; Zaslavsky, B. Y. Cooperativity between various types of polar solute-solvent interactions in aqueous media. J. Chromatogr. A 2015, 1408, 108-117.