(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%
Thermo Scientific Chemicals
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%
CAS: 22204-53-1 | C14H14O3 | 230.263 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFL09855.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
CAS22204-53-1
Health Hazard 1H301-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Qu-Ming Gu.; Ching-Shin Chen.; C.J Sih. A facile enzymatic resolution process for the preparation of (+)-Full-size image (<1 K)-2-(6-hethoxy-2-naphthyl)propionic acid (Naproxen).Tetrahedron Lett. 1986, 27 (16),1763-1766 .