4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
Thermo Scientific Chemicals
4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
CAS: 16588-69-5 | C8H3ClF3NO | 221.56 g/mol
Have Questions?
Catalog NumberQuantity
ALFL09850.065 g
Catalog number ALFL09850.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Chloro-2-(trifluoromethyl)phenyl isocyanate
CAS16588-69-5
Health Hazard 1H302+H312+H332-H315-H319-H334-H335
Health Hazard 2GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c