1,4-Phenylene diisothiocyanate, 98%
1,4-Phenylene diisothiocyanate, 98%
1,4-Phenylene diisothiocyanate, 98%
1,4-Phenylene diisothiocyanate, 98%
Thermo Scientific Chemicals
1,4-Phenylene diisothiocyanate, 98%
CAS: 4044-65-9 | C8H4N2S2 | 192.254 g/mol
Have Questions?
Catalog NumberQuantity
ALFL09756.065 g
Catalog number ALFL09756.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Phenylene diisothiocyanate
CAS4044-65-9
Health Hazard 1H301-H315-H319-H334-H335
Health Hazard 2GHS H Statement
H331-H334-H314-H318-H302-H312
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
View more
It is used in the treatment of hookworms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the treatment of hookworms.
Solubility
Hydrolyzes in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Thomas Deutschmann.; Helmut Ritter. Polymeric mesoions, 3. Synthesis of polymeric mesoionic 1,3-diazinium-olates via polymer analogous modification of a polythiourea from m-phenylenediamine and p-phenylene diisothiocyanate.Macromol. Rapid Commun. 1996, 17 (10),723-730 .