(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%
Thermo Scientific Chemicals
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%
CAS: 35132-20-8 | C14H18N2 | 214.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number ALFL09558.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine
CAS35132-20-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine is used as chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR. It is used in various catalyst systems for asymmetric reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine is used as chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR. It is used in various catalyst systems for asymmetric reactions.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat, air and oxidizing agents.
RUO – Research Use Only
General References:
  1. François Touchard; Patrick Gamez; Fabienne Fache; Marc Lemaire. Chiral thiourea as ligand for the asymmetric reduction of prochiral ketones. Chiral thiourea as ligand for the asymmetric reduction of prochiral ketones.Tetrahedron Letters.1997, 38 2275-2278.
  2. Chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR.
  3. In combination with various Lewis acids, forms a number of catalyst systems for asymmetric reactions including Diels-Alder, allylation and aldol reactions: Angew. Chem. Int. Ed., 30, 455 (1991). For use in enantioselective catalytic Diels-Alder reactions, see: J. Am. Chem. Soc., 111, 5493 (1989); Org. Synth. Coll., 9, 67 (1998).