Methyl 3,4-dihydroxybenzoate, 97%
Methyl 3,4-dihydroxybenzoate, 97%
Methyl 3,4-dihydroxybenzoate, 97%
Methyl 3,4-dihydroxybenzoate, 97%
Thermo Scientific Chemicals
Methyl 3,4-dihydroxybenzoate, 97%
CAS: 2150-43-8 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALFL09552.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl 3,4-dihydroxybenzoate
CAS2150-43-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c