10,12-Pentacosadiynoic acid, 98+%
10,12-Pentacosadiynoic acid, 98+%
10,12-Pentacosadiynoic acid, 98+%
10,12-Pentacosadiynoic acid, 98+%
Thermo Scientific Chemicals
10,12-Pentacosadiynoic acid, 98+%
CAS: 66990-32-7 | C25H42O2 | 374.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFL09313.14
Price (MYR)
4,232.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
4,232.00
EA
Add to cart
Specifications
Chemical Name or Material10,12-Pentacosadiynoic acid
CAS66990-32-7
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point62°C to 65°C
View more
10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.
Solubility
Slightly soluble in water.
Notes
Incompatible with reducing agents and oxidizing agents.
RUO – Research Use Only
General References:
  1. Yang, J. E.; Park, J. S.; Cho, E.; Jung, S.; Paik, S. R. Robust Polydiacetylene-Based Colorimetric Sensing Material Developed with Amyloid Fibrils of α-Synuclein. Langmuir 2015, 31 (5), 1802-1810.
  2. Hatta, E. Dislocation-Mediated Deformation in Solid Langmuir Monolayers: Plastic Bending and Tilt Boundary. Langmuir 2015, 31 (35), 9597-9601.