N,N-Dimethyldecanamide, 98%
N,N-Dimethyldecanamide, 98%
N,N-Dimethyldecanamide, 98%
N,N-Dimethyldecanamide, 98%
Thermo Scientific Chemicals
N,N-Dimethyldecanamide, 98%
CAS: 14433-76-2 | C12H25NO | 199.338 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08731.1425 g
Catalog number ALFL08731.14
Price (MYR)
2,307.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,307.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethyldecanamide
CAS14433-76-2
Recommended StorageAmbient temperatures
RTECS NumberHD6350000
EINECS Number238-405-1
View more
N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. W.J. Irwin; F.D. Sanderson; A. Li Wan Po. Percutaneous absorption of ibuprofen and naproxen: Effect of amide enhancers on transport through rat skin. International Journal of Pharmaceutics. 1990, 66 (1-3), 243-252.
  2. Yang-I Lin; S. A. LangJr.; Christina M. Seifert; R. G. Child; G. O. Morton; P. F. Fabio. Aldehyde Syntheses. Study of the preparation of 9,10-anthracenedicarboxaldehyde. J. Org. Chem. 1979, 44(25), 4701-4703.