N-Boc-D-phenylalanine, 98%
N-Boc-D-phenylalanine, 98%
N-Boc-D-phenylalanine, 98%
N-Boc-D-phenylalanine, 98%
Thermo Scientific Chemicals
N-Boc-D-phenylalanine, 98%
CAS: 18942-49-9 | C14H19NO4 | 265.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL08722.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Boc-D-phenylalanine
CAS18942-49-9
Melting Point85°C to 88°C
Recommended StorageKeep cold
TSCANo