Erucic acid, tech. 90%
Erucic acid, tech. 90%
Erucic acid, tech. 90%
Erucic acid, tech. 90%
Thermo Scientific Chemicals
Erucic acid, tech. 90%
CAS: 112-86-7 | C22H42O2 | 338.576 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08685.1850 g
Catalog number ALFL08685.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialErucic acid
CAS112-86-7
Melting Point29°C to 33°C
Recommended StorageAmbient temperatures
RTECS NumberJR1280000