4-Bromobenzenesulfonamide, 98%
4-Bromobenzenesulfonamide, 98%
4-Bromobenzenesulfonamide, 98%
4-Bromobenzenesulfonamide, 98%
Thermo Scientific Chemicals
4-Bromobenzenesulfonamide, 98%
CAS: 701-34-8 | C6H6BrNO2S | 236.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL08602.06
Price (MYR)
586.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
586.00
EA
Add to cart
Specifications
Chemical Name or Material4-Bromobenzenesulfonamide
CAS701-34-8
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c