3,5-Dimethylthiophenol, 97%
3,5-Dimethylthiophenol, 97%
3,5-Dimethylthiophenol, 97%
3,5-Dimethylthiophenol, 97%
3,5-Dimethylthiophenol, 97%
CAS: 38360-81-5 | C8H10S | 138.228 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08403.065 g
Catalog number ALFL08403.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3,5-Dimethylthiophenol
CAS38360-81-5
Health Hazard 1H227-H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c