2,3,4-Trimethylpentane, 98+%
2,3,4-Trimethylpentane, 98+%
2,3,4-Trimethylpentane, 98+%
2,3,4-Trimethylpentane, 98+%
Thermo Scientific Chemicals
2,3,4-Trimethylpentane, 98+%
CAS: 565-75-3 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL08176.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,3,4-Trimethylpentane
CAS565-75-3
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c