L-Noradrenaline, 98%
L-Noradrenaline, 98%
L-Noradrenaline, 98%
L-Noradrenaline, 98%
Thermo Scientific Chemicals
L-Noradrenaline, 98%
CAS: 51-41-2 | C8H11NO3 | 169.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08087.031 g
Catalog number ALFL08087.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Noradrenaline
CAS51-41-2
Health Hazard 1H300
Health Hazard 2GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodilatory shock states such as septic shock and neurogenic shock. Further, it is used in the preparation of1,2,3,4-tetrahydro-4,6,7-isoquinolinetriol hydrochloride. In addition to this, it is used in the treatment of hypotension, depression, schizophrenia and attention hyperactivity disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodil
RUO – Research Use Only
General References:
  1. Zhou, Q.; Kang, S. Z.; Li, X.; Wang, L.; Qin, L.; Mu, J. One-pot hydrothermal preparation of wurtzite CuGaS2 and its application as a photoluminescent probe for trace detection of L-noradrenaline. Colloids Surf., A 2015, 465, 124-129.
  2. Cui, L.; He, X. P.; Chen, G. R. Recent progress in quantum dot based sensors. RSC Adv. 2015, 5 (34), 26644-26653.