5-Methylisatin, 97%
5-Methylisatin, 97%
5-Methylisatin, 97%
Thermo Scientific Chemicals
5-Methylisatin, 97%
CAS: 608-05-9 | C9H7NO2 | 161.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08009.031 g
Catalog number ALFL08009.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Methylisatin
CAS608-05-9
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point∽180°C (decomposition)