1-Bromopentadecane, 97%
1-Bromopentadecane, 97%
1-Bromopentadecane, 97%
1-Bromopentadecane, 97%
Thermo Scientific Chemicals
1-Bromopentadecane, 97%
CAS: 629-72-1 | C15H31Br | 291.32 g/mol
Have Questions?
Catalog NumberQuantity
ALFL07618.0910 g
Catalog number ALFL07618.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromopentadecane
CAS629-72-1
Melting Point18°C to 19°C
Recommended StorageAmbient temperatures
Density1.005
View more
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
RUO – Research Use Only
M Roxana Sierra-Hernández; Heather C Allen. Incorporation and exclusion of long chain alkyl halides in fatty acid monolayers at the air-water interface.Langmuir.2010, 26 (24), 18806-18816.